Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000231751.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000417439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000426532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000431944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000431944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000431944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000431944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000431944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000431944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000431944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000431944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000443496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000462667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000462667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000462667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000462667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000478874.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000478874.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000478874.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000478874.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000478874.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000493056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000493056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000493056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000493056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000493056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000498301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000498301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000498301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
ENST00000498301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_001199149.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010949.hg.1 |
NM_002343.5 |
Refseq |