Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181672.2 | NP_858058.1 | 15 - 16 | 2210 | 82 | |
NM_181673.2 | NP_858059.1 | 15 - 16 | 2180 | 82 | ||
XM_017029907.1 | XP_016885396.1 | 13 - 14 | 1739 | 82 | ||
XM_017029908.1 | XP_016885397.1 | 13 - 14 | 5212 | 82 | ||
GenBank mRNA | AF223393.1 | - | 11 - 12 | 1847 | 82 | |
AK313530.1 | - | 11 - 12 | 2126 | 82 | ||
AL050366.1 | - | 10 - 11 | 3437 | 82 | ||
AL833085.2 | - | 15 - 16 | 2190 | 82 | ||
BC014434.1 | - | 15 - 16 | 2111 | 4865031 | 82 | |
BC038180.1 | - | 15 - 16 | 2143 | 5017795 | 82 | |
BX537844.1 | - | 15 - 16 | 2168 | 82 | ||
U77413.1 | - | 11 - 12 | 1859 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |