Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021515.hg.1 |
TC06001783.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021515.hg.1 |
ENST00000229810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021515.hg.1 |
ENST00000371175.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021515.hg.1 |
ENST00000418071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021515.hg.1 |
ENST00000539403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021515.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021516.hg.1 |
TC06001783.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021516.hg.1 |
ENST00000229810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021516.hg.1 |
ENST00000371175.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021516.hg.1 |
ENST00000418071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021516.hg.1 |
ENST00000539403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021516.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021517.hg.1 |
TC06001783.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021517.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011999.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182554.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182554.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182554.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182554.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182554.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182554.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182554.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182554.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182554.hg.1 |
TC0600011999.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600182553.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182553.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182553.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182553.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182553.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182553.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182553.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182553.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182553.hg.1 |
TC0600011999.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600182552.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182552.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182552.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182552.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182552.hg.1 |
ENST00000229810.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182552.hg.1 |
ENST00000371175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182552.hg.1 |
ENST00000618248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182552.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182552.hg.1 |
TC0600011999.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600182555.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182555.hg.1 |
NM_000324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182555.hg.1 |
TC0600011999.hg.1 |
Clariom D TC |