Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018703.hg.1 |
ENST00000298767.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018703.hg.1 |
ENST00000342368.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018703.hg.1 |
ENST00000372076.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018703.hg.1 |
NM_015045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018703.hg.1 |
TC10001480.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018702.hg.1 |
ENST00000263070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018702.hg.1 |
ENST00000298767.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018702.hg.1 |
ENST00000342368.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018702.hg.1 |
ENST00000372075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018702.hg.1 |
ENST00000372076.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018702.hg.1 |
NM_015045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018702.hg.1 |
TC10001480.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000176827.hg.1 |
ENST00000298767.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176827.hg.1 |
ENST00000618527.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176827.hg.1 |
NM_015045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176827.hg.1 |
ENST00000298767.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176827.hg.1 |
ENST00000618527.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176827.hg.1 |
NM_015045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176827.hg.1 |
TC1000011299.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000176826.hg.1 |
ENST00000263070.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176826.hg.1 |
ENST00000298767.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176826.hg.1 |
ENST00000372075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176826.hg.1 |
ENST00000618527.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176826.hg.1 |
NM_015045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176826.hg.1 |
ENST00000263070.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176826.hg.1 |
ENST00000298767.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176826.hg.1 |
ENST00000372075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176826.hg.1 |
ENST00000618527.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176826.hg.1 |
NM_015045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176826.hg.1 |
TC1000011299.hg.1 |
Clariom D TC |