Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038686.hg.1 |
TC01002493.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038686.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038686.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038687.hg.1 |
TC01002493.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038687.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038687.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241798.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241798.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241798.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241798.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241798.hg.1 |
TC0100018432.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
ENST00000357214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018432.hg.1 |
NM_005066.2 |
Refseq |