Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005068.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005068.hg.1 |
NM_178455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005068.hg.1 |
TC20000353.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007498.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007498.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007498.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007498.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007498.hg.1 |
NM_178455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007498.hg.1 |
NM_178455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007498.hg.1 |
NM_178455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005069.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005069.hg.1 |
NM_178455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005069.hg.1 |
TC20000353.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000152214.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152214.hg.1 |
NM_178455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152214.hg.1 |
TC2000007498.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000152214.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152214.hg.1 |
NM_178455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007498.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007498.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007498.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007498.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007498.hg.1 |
NM_178455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007498.hg.1 |
NM_178455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007498.hg.1 |
NM_178455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152213.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152213.hg.1 |
NM_178455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152213.hg.1 |
TC2000007498.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000152213.hg.1 |
ENST00000279058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152213.hg.1 |
NM_178455.2 |
Refseq |