Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060599.hg.1 |
TC01004003.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060599.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060599.hg.1 |
ENST00000366595.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060599.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100234682.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100234682.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100234682.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100234682.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100234682.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100234682.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100234682.hg.1 |
TC0100017844.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000264187.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
ENST00000366595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017844.hg.1 |
NM_002508.2 |
Refseq |