Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000474306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000474306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000474306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000489355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000489355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000489355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006252.hg.1 |
TC09000560.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006252.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006252.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006252.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156430.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156430.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156430.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156430.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156430.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156430.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156430.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156430.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156430.hg.1 |
TC0900008441.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000374279.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000474306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000474306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000474306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000489355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000489355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000489355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000490110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
ENST00000495085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008441.hg.1 |
NM_003358.2 |
Refseq |