Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031092.hg.1 |
TC07002032.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031092.hg.1 |
ENST00000275838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031092.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031092.hg.1 |
ENST00000422024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031092.hg.1 |
ENST00000434669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031092.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031092.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031090.hg.1 |
TC07002032.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031090.hg.1 |
ENST00000275838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031090.hg.1 |
ENST00000377867.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031090.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031090.hg.1 |
ENST00000422024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031090.hg.1 |
ENST00000434669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031090.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031090.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031090.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031095.hg.1 |
TC07002032.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031095.hg.1 |
ENST00000275838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031095.hg.1 |
ENST00000422024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031095.hg.1 |
ENST00000434669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031095.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031095.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031093.hg.1 |
TC07002032.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031093.hg.1 |
ENST00000275838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031093.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031093.hg.1 |
ENST00000422024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031093.hg.1 |
ENST00000434669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031093.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031093.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031094.hg.1 |
TC07002032.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031094.hg.1 |
ENST00000275838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031094.hg.1 |
ENST00000422024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031094.hg.1 |
ENST00000434669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031094.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031094.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194211.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194211.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194211.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194211.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194211.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194211.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194211.hg.1 |
TC0700013063.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013063.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194208.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194208.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194208.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194208.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194208.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194208.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194208.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194208.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194208.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194208.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194208.hg.1 |
TC0700013063.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700194210.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194210.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194210.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194210.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194210.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194210.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194210.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194210.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194210.hg.1 |
TC0700013063.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
ENST00000377867.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
NM_080871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194206.hg.1 |
TC0700013063.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700194209.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194209.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194209.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194209.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194209.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194209.hg.1 |
ENST00000420175.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194209.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194209.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194209.hg.1 |
TC0700013063.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700194212.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194212.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194212.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194212.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194212.hg.1 |
ENST00000275838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194212.hg.1 |
ENST00000415615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194212.hg.1 |
NM_001142459.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194212.hg.1 |
NM_001142460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194212.hg.1 |
TC0700013063.hg.1 |
Clariom D TC |