Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286613.1 | NP_001273542.1 | 8 - 9 | 1535 | 136 | |
NM_022765.3 | NP_073602.3 | 8 - 9 | 1499 | 136 | ||
GenBank mRNA | AB048948.1 | - | 8 - 9 | 1499 | 136 | |
AK021999.1 | - | 7 - 8 | 1242 | 136 | ||
AK025392.1 | - | 8 - 9 | 1266 | 136 | ||
AK160384.1 | - | 5 - 6 | 2095 | 136 | ||
AK225049.1 | - | 8 - 9 | 1256 | 136 | ||
AK296284.1 | - | 8 - 9 | 1538 | 136 | ||
BC042144.1 | - | 8 - 9 | 1395 | 5929774 | 136 | |
BC052983.1 | - | 7 - 8 | 1212 | 5756097 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |