Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005734.hg.1 |
ENST00000309156.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005734.hg.1 |
ENST00000309185.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005734.hg.1 |
ENST00000394204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005734.hg.1 |
ENST00000394209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005734.hg.1 |
ENST00000394211.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005734.hg.1 |
ENST00000445327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005734.hg.1 |
NM_001030002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005734.hg.1 |
NM_001242442.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005734.hg.1 |
NM_001242443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005734.hg.1 |
NM_005310.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005734.hg.1 |
TC17000481.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
ENST00000309156.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
ENST00000394204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
ENST00000394209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
ENST00000394211.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
ENST00000445327.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
NM_001030002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
NM_001242442.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
NM_001242443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
NM_005310.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
ENST00000309156.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
ENST00000394204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
ENST00000394209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
ENST00000394211.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
ENST00000445327.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
NM_001030002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
NM_001242442.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
NM_001242443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
NM_005310.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156292.hg.1 |
TC1700007757.hg.1 |
Clariom D TC |