Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002111.8 | NP_002102.4 | 20 - 21 | 2851 | 110 | |
GenBank mRNA | AB016794.1 | - | 20 - 21 | 2851 | 110 | |
AB209506.1 | - | 7 - 8 | 2799 | 110 | ||
L12392.1 | - | 20 - 21 | 3021 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |