Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017374.hg.1 |
TC09001395.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017374.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017374.hg.1 |
ENST00000354801.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017374.hg.1 |
ENST00000375172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017374.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375163.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375163.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375163.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375163.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375163.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375163.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000375173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000484708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000484708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
ENST00000484708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017369.hg.1 |
TC09001395.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017369.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017369.hg.1 |
ENST00000354801.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017369.hg.1 |
ENST00000375165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017369.hg.1 |
ENST00000375172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017369.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017376.hg.1 |
TC09001395.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017376.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017376.hg.1 |
ENST00000354801.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017376.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017373.hg.1 |
TC09001395.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017373.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017373.hg.1 |
ENST00000354801.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017373.hg.1 |
ENST00000375172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017373.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017377.hg.1 |
TC09001395.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017377.hg.1 |
ENST00000354801.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017377.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174608.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174608.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174608.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174608.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174608.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174608.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174608.hg.1 |
TC0900010946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375163.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375163.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375163.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375163.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375163.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375163.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000375173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000484708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000484708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
ENST00000484708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010946.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174604.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174604.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174604.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174604.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174604.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174604.hg.1 |
ENST00000375165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174604.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174604.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174604.hg.1 |
TC0900010946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900174611.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174611.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174611.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174611.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174611.hg.1 |
TC0900010946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900174612.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174612.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174612.hg.1 |
TC0900010946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900174607.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174607.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174607.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174607.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174607.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174607.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174607.hg.1 |
TC0900010946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900174609.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174609.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174609.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174609.hg.1 |
ENST00000341170.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174609.hg.1 |
ENST00000375172.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174609.hg.1 |
NM_139246.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174609.hg.1 |
TC0900010946.hg.1 |
Clariom D TC |