Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007553.2 | NP_001007554.1 | 5 - 6 | 907 | 82 | |
NM_001242891.1 | NP_001229820.1 | 6 - 7 | 1054 | 82 | ||
NM_001242892.1 | NP_001229821.1 | 4 - 5 | 520 | 82 | ||
GenBank mRNA | AB020692.1 | - | 5 - 6 | 821 | 82 | |
AK290146.1 | - | 4 - 5 | 452 | 82 | ||
CR749378.1 | - | 5 - 6 | 911 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |