Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBL1Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134258.1 | NP_599020.1 | 2 - 3 | 382 | 125 | |
GenBank mRNA | AF332221.1 | - | 2 - 3 | 382 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |