Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015695.2 | NP_056510.2 | 10 - 11 | 3402 | 71 | |
XM_005249010.2 | XP_005249067.1 | 10 - 11 | 3284 | 71 | ||
XM_005249011.3 | XP_005249068.1 | 10 - 11 | 3408 | 71 | ||
XM_011514489.1 | XP_011512791.1 | 10 - 11 | 3306 | 71 | ||
XM_011514490.1 | XP_011512792.1 | 9 - 10 | 3204 | 71 | ||
XM_011514491.1 | XP_011512793.1 | 9 - 10 | 3204 | 71 | ||
XM_017010743.1 | XP_016866232.1 | 7 - 8 | 1520 | 71 | ||
GenBank mRNA | AB033112.1 | - | 10 - 11 | 3402 | 71 | |
AK025134.1 | - | 3 - 4 | 462 | 71 | ||
AK074362.1 | - | 7 - 8 | 1540 | 71 | ||
BC117387.1 | - | 7 - 8 | 2399 | 40125938 | 71 | |
BC143917.1 | - | 7 - 8 | 2399 | 9052435 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |