Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000339717.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000357641.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000441730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000446974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000446974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000446974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000446974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000446974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000449261.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000454960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000454960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000454960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000454960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000454960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000454960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000527657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000527657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000527657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000532538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
ENST00000534694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007832.hg.1 |
NM_015695.2 |
Refseq |