Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004369.3 | NP_004360.2 | 34 - 35 | 7448 | 84 | |
NM_057166.4 | NP_476507.3 | 31 - 32 | 5627 | 84 | ||
NM_057167.3 | NP_476508.2 | 33 - 34 | 6830 | 84 | ||
XM_005246065.1 | XP_005246122.1 | 33 - 34 | 6805 | 84 | ||
XM_005246066.1 | XP_005246123.1 | 31 - 32 | 5951 | 84 | ||
XM_006712253.1 | XP_006712316.1 | 34 - 35 | 6904 | 84 | ||
XM_011510574.1 | XP_011508876.1 | 34 - 35 | 7402 | 84 | ||
XM_017003303.1 | XP_016858792.1 | 39 - 40 | 7455 | 84 | ||
XM_017003304.1 | XP_016858793.1 | 36 - 37 | 5667 | 84 | ||
GenBank mRNA | AB209636.1 | - | 25 - 26 | 2739 | 84 | |
BC033174.1 | - | 11 - 12 | 571 | 84 | ||
BC144595.1 | - | 31 - 32 | 5568 | 9053125 | 84 | |
BC150625.1 | - | 31 - 32 | 5568 | 9056995 | 84 | |
BC171790.1 | - | 31 - 32 | 5568 | 9054444 | 84 | |
BX641155.1 | - | 11 - 12 | 1417 | 84 | ||
X52022.1 | - | 34 - 35 | 7418 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons |