Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017431.2 | NP_059127.2 | 11 - 12 | 1225 | 61 | |
XM_017004343.1 | XP_016859832.1 | 11 - 12 | 1239 | 61 | ||
GenBank mRNA | AF214519.1 | - | 9 - 10 | 1131 | 61 | |
AJ249977.1 | - | 11 - 12 | 1227 | 61 | ||
AK300078.1 | - | 12 - 13 | 1191 | 61 | ||
BC098102.1 | - | 11 - 12 | 1225 | 40005880 | 61 | |
BC098255.1 | - | 11 - 12 | 1225 | 40005883 | 61 | |
BC098277.1 | - | 11 - 12 | 1225 | 40005886 | 61 | |
BC098306.1 | - | 11 - 12 | 1225 | 40005888 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |