Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012773.hg.1 |
ENST00000244043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012773.hg.1 |
ENST00000478764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012773.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012773.hg.1 |
TC20000927.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012774.hg.1 |
ENST00000244043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012774.hg.1 |
ENST00000478764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012774.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012774.hg.1 |
TC20000927.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165692.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165692.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165692.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165692.hg.1 |
TC2000009388.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000165692.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165692.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165692.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009388.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165693.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165693.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165693.hg.1 |
NM_000961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165693.hg.1 |
TC2000009388.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000165693.hg.1 |
ENST00000244043.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165693.hg.1 |
ENST00000478971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165693.hg.1 |
NM_000961.3 |
Refseq |