Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008537.hg.1 |
TC07000587.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008537.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008537.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000466102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000466102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000466102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
ENST00000466102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008536.hg.1 |
TC07000587.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008536.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008536.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158994.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158994.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158994.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158994.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158994.hg.1 |
TC0700008465.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000466102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000466102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000466102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
ENST00000466102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008465.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158993.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158993.hg.1 |
ENST00000466102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158993.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158993.hg.1 |
ENST00000265634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158993.hg.1 |
ENST00000466102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158993.hg.1 |
NM_002523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158993.hg.1 |
TC0700008465.hg.1 |
Clariom D TC |