Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOPEY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199942.1 | NP_001186871.1 | 28 - 29 | 6133 | 257 | |
NM_015018.3 | NP_055833.2 | 28 - 29 | 6160 | 257 | ||
XM_011535619.2 | XP_011533921.1 | 27 - 28 | 6125 | 257 | ||
XM_017010559.1 | XP_016866048.1 | 28 - 29 | 6155 | 257 | ||
XM_017010560.1 | XP_016866049.1 | 27 - 28 | 6083 | 257 | ||
XM_017010561.1 | XP_016866050.1 | 28 - 29 | 6107 | 257 | ||
XM_017010562.1 | XP_016866051.1 | 27 - 28 | 6056 | 257 | ||
XM_017010563.1 | XP_016866052.1 | 28 - 29 | 6134 | 257 | ||
XM_017010564.1 | XP_016866053.1 | 28 - 29 | 6134 | 257 | ||
XM_017010565.1 | XP_016866054.1 | 28 - 29 | 6128 | 257 | ||
XM_017010566.1 | XP_016866055.1 | 28 - 29 | 6128 | 257 | ||
XM_017010567.1 | XP_016866056.1 | 27 - 28 | 6056 | 257 | ||
XM_017010568.1 | XP_016866057.1 | 27 - 28 | 6125 | 257 | ||
XM_017010569.1 | XP_016866058.1 | 26 - 27 | 6042 | 257 | ||
XM_017010570.1 | XP_016866059.1 | 27 - 28 | 6056 | 257 | ||
XM_017010571.1 | XP_016866060.1 | 27 - 28 | 6050 | 152 | ||
XM_017010572.1 | XP_016866061.1 | 28 - 29 | 6155 | 257 | ||
XM_017010573.1 | XP_016866062.1 | 28 - 29 | 6155 | 257 | ||
XM_017010574.1 | XP_016866063.1 | 28 - 29 | 6155 | 257 | ||
GenBank mRNA | AB029040.2 | - | 11 - 12 | 3181 | 257 | |
AK027030.1 | - | 8 - 9 | 1927 | 257 | ||
BC144544.1 | - | 28 - 29 | 6031 | 9053071 | 257 | |
BC150619.1 | - | 28 - 29 | 6031 | 9056989 | 257 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |