Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000484860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000484860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000491469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000491469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000491469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000491469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
ENST00000491469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005893.hg.1 |
ENST00000262593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005893.hg.1 |
ENST00000395939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005893.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005893.hg.1 |
TC20000424.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000484860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000484860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000491469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000491469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000491469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000491469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
ENST00000491469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_001294161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007797.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000154131.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154131.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154131.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000154131.hg.1 |
NM_177959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000154131.hg.1 |
TC2000007797.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000154131.hg.1 |
ENST00000262593.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154131.hg.1 |
ENST00000395939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154131.hg.1 |
NM_018431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000154131.hg.1 |
NM_177959.2 |
Refseq |