Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020679.hg.1 |
TC06001730.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020679.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020679.hg.1 |
ENST00000372949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020679.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020678.hg.1 |
TC06001730.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020678.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020678.hg.1 |
ENST00000372949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020678.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020680.hg.1 |
TC06001730.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020680.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020680.hg.1 |
ENST00000372949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020680.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020681.hg.1 |
TC06001730.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020681.hg.1 |
ENST00000372949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020681.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181199.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181199.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181199.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181199.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181199.hg.1 |
TC0600011821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011821.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181197.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181197.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181197.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181197.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181197.hg.1 |
TC0600011821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600181196.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181196.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181196.hg.1 |
ENST00000230361.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181196.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181196.hg.1 |
TC0600011821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600181200.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181200.hg.1 |
NM_002098.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181200.hg.1 |
TC0600011821.hg.1 |
Clariom D TC |