Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012355.hg.1 |
TC0X000915.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012355.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012355.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012355.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012355.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009227.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163432.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163432.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163432.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163432.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163432.hg.1 |
ENST00000379494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163432.hg.1 |
ENST00000494525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163432.hg.1 |
NM_014332.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163432.hg.1 |
NR_045617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163432.hg.1 |
TC0X00009227.hg.1 |
Clariom D TC |