Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000468041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000468041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
ENST00000468041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002056.hg.1 |
TC09000185.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002056.hg.1 |
ENST00000361226.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002056.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002056.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002056.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000468041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000468041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
ENST00000468041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007082.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148104.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148104.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148104.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148104.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148104.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148104.hg.1 |
ENST00000361226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148104.hg.1 |
ENST00000455600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148104.hg.1 |
NM_001135999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148104.hg.1 |
NM_014806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148104.hg.1 |
NR_052015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148104.hg.1 |
TC0900007082.hg.1 |
Clariom D TC |