Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000197.hg.1 |
ENST00000263620.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000197.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000197.hg.1 |
TC19000023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000198.hg.1 |
ENST00000263620.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000198.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000198.hg.1 |
TC19000023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000592216.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000592216.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000199.hg.1 |
ENST00000263620.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000199.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000199.hg.1 |
TC19000023.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900144605.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144605.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144605.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144605.hg.1 |
TC1900006499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144605.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144605.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144605.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144607.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144607.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144607.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144607.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144607.hg.1 |
TC1900006499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144607.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144607.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144607.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144607.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144606.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144606.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144606.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144606.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144606.hg.1 |
TC1900006499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144606.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144606.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144606.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144606.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144600.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144600.hg.1 |
ENST00000585895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144600.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144600.hg.1 |
TC1900006499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144600.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144600.hg.1 |
ENST00000585895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144600.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000592216.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000592216.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144598.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144598.hg.1 |
ENST00000585895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144598.hg.1 |
ENST00000592216.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144598.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144598.hg.1 |
TC1900006499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144598.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144598.hg.1 |
ENST00000585895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144598.hg.1 |
ENST00000592216.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144598.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144603.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144603.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144603.hg.1 |
TC1900006499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144603.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144603.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144604.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144604.hg.1 |
NM_005224.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144604.hg.1 |
TC1900006499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144604.hg.1 |
ENST00000263620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144604.hg.1 |
NM_005224.2 |
Refseq |