Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027369.hg.1 |
TC01001748.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027369.hg.1 |
ENST00000009105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027369.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027369.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027369.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000423146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011451.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184327.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184327.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184327.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184327.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184327.hg.1 |
ENST00000009105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184327.hg.1 |
ENST00000361322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184327.hg.1 |
ENST00000494990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184327.hg.1 |
NM_020439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184327.hg.1 |
TC0100011451.hg.1 |
Clariom D TC |