Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043855.hg.1 |
TC01002819.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043855.hg.1 |
ENST00000370630.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043855.hg.1 |
ENST00000477677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043855.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014755.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209866.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209866.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209866.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209866.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209866.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209866.hg.1 |
ENST00000370625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209866.hg.1 |
ENST00000370630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209866.hg.1 |
ENST00000465118.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209866.hg.1 |
ENST00000477677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209866.hg.1 |
NM_004388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209866.hg.1 |
TC0100014755.hg.1 |
Clariom D TC |