Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000487208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000487208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000487208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000487208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000491503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
ENST00000491503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057775.hg.1 |
TC01003768.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057775.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057775.hg.1 |
ENST00000487208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057775.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229752.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229752.hg.1 |
ENST00000487208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229752.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229752.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229752.hg.1 |
ENST00000487208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229752.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229752.hg.1 |
TC0100017113.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000356495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000487208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000487208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000487208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000487208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000491503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
ENST00000491503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017113.hg.1 |
NM_002644.3 |
Refseq |