Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011099.hg.1 |
TC02000715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011099.hg.1 |
ENST00000259205.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011099.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000447128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000447128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000447128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000447128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
ENST00000447128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011100.hg.1 |
TC02000715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011100.hg.1 |
ENST00000259205.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011100.hg.1 |
ENST00000376489.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011100.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000447128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000447128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000447128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000447128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
ENST00000447128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008945.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162464.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162464.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162464.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162464.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162464.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162464.hg.1 |
ENST00000376489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162464.hg.1 |
NM_001278568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162464.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162464.hg.1 |
TC0200008945.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200162463.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162463.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162463.hg.1 |
ENST00000259205.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162463.hg.1 |
NM_019618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162463.hg.1 |
TC0200008945.hg.1 |
Clariom D TC |