Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020247.4 | NP_064632.2 | 10 - 11 | 1426 | 98 | |
XM_005273201.1 | XP_005273258.1 | 10 - 11 | 1322 | 98 | ||
XM_011544238.1 | XP_011542540.1 | 11 - 12 | 1518 | 98 | ||
XM_011544239.2 | XP_011542541.1 | 12 - 13 | 1541 | 98 | ||
XM_011544240.2 | XP_011542542.1 | 10 - 11 | 1335 | 98 | ||
XM_011544241.2 | XP_011542543.1 | 10 - 11 | 1297 | 98 | ||
XM_017001852.1 | XP_016857341.1 | 10 - 11 | 1516 | 98 | ||
GenBank mRNA | AB073905.1 | - | 10 - 11 | 1349 | 98 | |
AJ278126.1 | - | 6 - 7 | 644 | 98 | ||
AK074693.1 | - | 10 - 11 | 1369 | 98 | ||
AK090494.1 | - | 10 - 11 | 1163 | 98 | ||
AK092784.1 | - | 10 - 11 | 1149 | 98 | ||
AK297735.1 | - | 7 - 8 | 705 | 98 | ||
AK300537.1 | - | 6 - 7 | 98 | |||
AK301012.1 | - | 12 - 13 | 1347 | 98 | ||
AL137511.1 | - | 5 - 6 | 98 | |||
BC005171.2 | - | 10 - 11 | 1375 | 3604948 | 98 | |
BX648860.1 | - | 10 - 11 | 1247 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |