Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000436738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
ENST00000436738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024203.hg.1 |
TC04001755.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024203.hg.1 |
ENST00000274093.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024203.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024203.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024203.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185448.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185448.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185448.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185448.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185448.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185448.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185448.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185448.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185448.hg.1 |
TC0400012462.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000274093.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000340217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000436738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
ENST00000436738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_001042543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012462.hg.1 |
NM_006529.3 |
Refseq |