Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024360.hg.1 |
ENST00000327268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024360.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024360.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024360.hg.1 |
TC17001820.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000582867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000582867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000582867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000582867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000582867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000582867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000582867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000582867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000582867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000582867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011558.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
ENST00000327268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
ENST00000580666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
ENST00000583477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
ENST00000620436.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
NM_001174166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
NM_004694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700193490.hg.1 |
TC1700011558.hg.1 |
Clariom D TC |