Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021171.hg.1 |
TC04001494.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021171.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021171.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021170.hg.1 |
TC04001494.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021170.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021170.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000510903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000510903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000510903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000515089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000515089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000515089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000515089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
ENST00000515089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180241.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180241.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180241.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180241.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180241.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180241.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180241.hg.1 |
TC0400011685.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000510903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000510903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000510903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000515089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000515089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000515089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000515089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
ENST00000515089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011685.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180242.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180242.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180242.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180242.hg.1 |
ENST00000296498.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180242.hg.1 |
ENST00000503043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180242.hg.1 |
NM_003619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180242.hg.1 |
TC0400011685.hg.1 |
Clariom D TC |