Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000309951.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000379923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
ENST00000541043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_001278352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006937.hg.1 |
NM_002197.2 |
Refseq |