Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016256.hg.1 |
ENST00000333430.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016256.hg.1 |
ENST00000439028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016256.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016256.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016256.hg.1 |
TC19001167.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016254.hg.1 |
ENST00000333430.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016254.hg.1 |
ENST00000359134.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016254.hg.1 |
ENST00000439028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016254.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016254.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016254.hg.1 |
TC19001167.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016258.hg.1 |
ENST00000333430.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016258.hg.1 |
ENST00000359134.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016258.hg.1 |
ENST00000439028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016258.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016258.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016258.hg.1 |
TC19001167.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016264.hg.1 |
ENST00000439028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016264.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016264.hg.1 |
TC19001167.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000590601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000590601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000590601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178009.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178009.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178009.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178009.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178009.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178009.hg.1 |
TC1900009633.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900178009.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178009.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178009.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178009.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178009.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178018.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178018.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178018.hg.1 |
TC1900009633.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900178018.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178018.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178019.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178019.hg.1 |
TC1900009633.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900178019.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178008.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178008.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178008.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178008.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178008.hg.1 |
TC1900009633.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900178008.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178008.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178008.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178008.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
ENST00000590601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
TC1900009633.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
ENST00000590601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900178010.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000333430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000439028.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000590601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000590601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000590601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
ENST00000617721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_001166215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009633.hg.1 |
NM_030760.4 |
Refseq |