Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143831.2 | NP_001137303.1 | 8 - 9 | 2922 | 57 | |
XM_006718828.3 | XP_006718891.1 | 9 - 10 | 3118 | 57 | ||
XM_011542792.1 | XP_011541094.1 | 9 - 10 | 3120 | 57 | ||
GenBank mRNA | D28539.1 | - | 8 - 9 | 2872 | 57 | |
S64316.1 | - | 2 - 3 | 145 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Homo sapiens Probe spans exons |