Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_017584_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8074076 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8074073 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8074080 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8074072 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8074079 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8074081 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8074075 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8074071 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8074078 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8074074 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8074077 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8074070 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
224179_PM_s_at |
- |
- |
Affymetrix Human Genome U219 Array |
11745562_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11753272_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11745562_PM_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11753272_PM_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11742739_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11742739_PM_a_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008097.hg.1 |
ENST00000216075.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008097.hg.1 |
ENST00000395732.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008097.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008097.hg.1 |
TC22000427.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008103.hg.1 |
ENST00000395732.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008103.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008103.hg.1 |
TC22000427.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008100.hg.1 |
ENST00000216075.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008100.hg.1 |
ENST00000395732.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008100.hg.1 |
ENST00000395733.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008100.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008100.hg.1 |
TC22000427.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008099.hg.1 |
ENST00000216075.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008099.hg.1 |
ENST00000395732.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008099.hg.1 |
ENST00000395733.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008099.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008099.hg.1 |
TC22000427.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200155469.hg.1 |
TC2200007809.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200155469.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155469.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155469.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155469.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155469.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155469.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155469.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155469.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155476.hg.1 |
TC2200007809.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200155476.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155476.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155476.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155476.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155473.hg.1 |
TC2200007809.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200155473.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155473.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155473.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155473.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155473.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155473.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155473.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155473.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
Human Genome U133 Plus 2.0 Array |
224179_PM_s_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
224179_s_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_017584_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224179_s_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224179_s_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224179_s_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224179_s_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224179_s_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224179_s_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224179_s_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224179_s_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224179_s_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224179_s_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224179_s_at2 |
- |
- |
Human Genome U95 Set |
78420_at |
- |
- |
Human Genome U95 Set |
78420_PM_at |
- |
- |
Human Genome U133 Set |
224179_PM_s_at |
- |
- |
Human Genome U133 Set |
224179_s_at |
- |
- |