Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323250.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323250.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323250.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323250.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323250.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000584122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
ENST00000584122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18000059.hg.1 |
ENST00000323250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18000059.hg.1 |
ENST00000323274.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18000059.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18000059.hg.1 |
TC18000006.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800144213.hg.1 |
ENST00000323250.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800144213.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800144213.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800144213.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800144213.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800144213.hg.1 |
ENST00000323250.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800144213.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800144213.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800144213.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800144213.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800144213.hg.1 |
TC1800006448.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323224.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323250.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323250.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323250.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323250.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323250.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000323274.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000579128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000581920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000584122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
ENST00000584122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006448.hg.1 |
NM_001071.2 |
Refseq |