Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004063.hg.1 |
ENST00000300935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004063.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004063.hg.1 |
TC19000317.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004060.hg.1 |
ENST00000300935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004060.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004060.hg.1 |
TC19000317.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009331.hg.1 |
ENST00000631259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000588105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000588105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000592971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000592971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000592971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
ENST00000592971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004061.hg.1 |
ENST00000300935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004061.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004061.hg.1 |
TC19000317.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004062.hg.1 |
ENST00000300935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004062.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004062.hg.1 |
TC19000317.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900199756.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199756.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199756.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199756.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199756.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199756.hg.1 |
TC1900011682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900199756.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199756.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199756.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199756.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199756.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199757.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199757.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199757.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199757.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199757.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199757.hg.1 |
TC1900011682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900199757.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199757.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199757.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199757.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199757.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199749.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199749.hg.1 |
TC1900011682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900199749.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009331.hg.1 |
ENST00000631259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199748.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199748.hg.1 |
TC1900011682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900199748.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
ENST00000588105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
TC1900011682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
ENST00000588105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199754.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000586682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000587156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000588105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000588105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000589697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000590899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000592971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000592971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000592971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
ENST00000592971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011682.hg.1 |
NM_005370.4 |
Refseq |