Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017892.3 | NP_060362.3 | 9 - 10 | 677 | 144 | |
XM_005246676.4 | XP_005246733.3 | 9 - 10 | 1197 | 144 | ||
XM_005246677.4 | XP_005246734.2 | 9 - 10 | 673 | 144 | ||
XM_011511446.2 | XP_011509748.1 | 8 - 9 | 1143 | 90 | ||
XM_011511447.2 | XP_011509749.1 | 9 - 10 | 1197 | 144 | ||
XM_011511448.2 | XP_011509750.1 | 8 - 9 | 619 | 90 | ||
XM_011511449.2 | XP_011509751.1 | 11 - 12 | 1027 | 144 | ||
XM_017004453.1 | XP_016859942.1 | 9 - 10 | 1197 | 144 | ||
XM_017004454.1 | XP_016859943.1 | 8 - 9 | 1143 | 90 | ||
XM_017004455.1 | XP_016859944.1 | 8 - 9 | 619 | 90 | ||
XM_017004456.1 | XP_016859945.1 | 10 - 11 | 863 | 144 | ||
XM_017004457.1 | XP_016859946.1 | 11 - 12 | 1026 | 144 | ||
XM_017004458.1 | XP_016859947.1 | 9 - 10 | 773 | 90 | ||
XM_017004459.1 | XP_016859948.1 | 9 - 10 | 882 | 90 | ||
XM_017004460.1 | XP_016859949.1 | 9 - 10 | 810 | 90 | ||
XM_017004461.1 | XP_016859950.1 | 9 - 10 | 882 | 90 | ||
GenBank mRNA | AF049523.1 | - | 9 - 10 | 662 | 144 | |
AF049528.1 | - | 9 - 10 | 749 | 144 | ||
AK024810.1 | - | 10 - 11 | 996 | 144 | ||
AK298547.1 | - | 6 - 7 | 514 | 144 | ||
BC000273.1 | - | 9 - 10 | 640 | 144 | ||
BC011788.2 | - | 9 - 10 | 661 | 144 | ||
BC029414.1 | - | 9 - 10 | 677 | 144 | ||
U70667.1 | - | 8 - 9 | 661 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |