Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017968.hg.1 |
ENST00000253669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017968.hg.1 |
ENST00000448593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017968.hg.1 |
NM_001011699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017968.hg.1 |
NM_033417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017968.hg.1 |
TC19001269.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
ENST00000253669.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
ENST00000448593.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
ENST00000593360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
ENST00000598517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
ENST00000601564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
NM_001011699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
NM_033417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
TC1900009943.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
ENST00000253669.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
ENST00000448593.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
ENST00000593360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
ENST00000598517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
ENST00000601564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
NM_001011699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900181666.hg.1 |
NM_033417.1 |
Refseq |