Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTBN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016642.3 | NP_057726.4 | 65 - 66 | 11063 | 63 | |
XM_017022299.1 | XP_016877788.1 | 63 - 64 | 11041 | 63 | ||
XM_017022300.1 | XP_016877789.1 | 64 - 65 | 10921 | 63 | ||
XM_017022301.1 | XP_016877790.1 | 59 - 60 | 10492 | 63 | ||
XM_017022302.1 | XP_016877791.1 | 51 - 52 | 8290 | 63 | ||
GenBank mRNA | AF233523.1 | - | 65 - 66 | 11063 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons |