Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002389.hg.1 |
ENST00000221214.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002389.hg.1 |
ENST00000260357.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002389.hg.1 |
ENST00000456763.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002389.hg.1 |
ENST00000457542.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002389.hg.1 |
ENST00000514566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002389.hg.1 |
NM_001128608.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002389.hg.1 |
NM_014994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002389.hg.1 |
TC15000313.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
ENST00000456763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
ENST00000457542.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
ENST00000505061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
ENST00000505373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
ENST00000512970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
ENST00000514566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
NM_001128608.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
NM_001265611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
NM_014994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
NR_049761.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
NR_049762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
ENST00000456763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
ENST00000457542.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
ENST00000505061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
ENST00000505373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
ENST00000512970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
ENST00000514566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
NM_001128608.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
NM_001265611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
NM_014994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
NR_049761.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
NR_049762.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149201.hg.1 |
TC1500007012.hg.1 |
Clariom D TC |