Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000460714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000460714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009909.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009909.hg.1 |
ENST00000537906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009909.hg.1 |
ENST00000541666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009909.hg.1 |
ENST00000542051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009909.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009909.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009909.hg.1 |
TC10000824.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000161213.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161213.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161213.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161213.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161213.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161213.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161213.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161213.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161213.hg.1 |
TC1000008951.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000351270.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000460714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000460714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
ENST00000542051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_001177660.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008951.hg.1 |
NM_004132.4 |
Refseq |