Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000472597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000472597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000472597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000472597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000472903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000472903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000472903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000472903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000472903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000509221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000509221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
ENST00000509221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005311.hg.1 |
TC04000389.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005311.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005311.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153191.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153191.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153191.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153191.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153191.hg.1 |
TC0400007785.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000396073.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000472597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000472597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000472597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000472597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000472903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000472903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000472903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000472903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000472903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000509221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000509221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
ENST00000509221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007785.hg.1 |
NM_031889.2 |
Refseq |