Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003565.hg.1 |
TC09000333.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003565.hg.1 |
ENST00000297784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003565.hg.1 |
ENST00000340019.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003565.hg.1 |
ENST00000396237.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003565.hg.1 |
ENST00000486417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003565.hg.1 |
ENST00000537917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003565.hg.1 |
ENST00000538054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003565.hg.1 |
ENST00000542143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003565.hg.1 |
NM_138691.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151199.hg.1 |
ENST00000297784.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151199.hg.1 |
ENST00000340019.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151199.hg.1 |
ENST00000486417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151199.hg.1 |
NM_138691.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151199.hg.1 |
ENST00000297784.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151199.hg.1 |
ENST00000340019.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151199.hg.1 |
ENST00000486417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151199.hg.1 |
NM_138691.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151199.hg.1 |
TC0900007563.hg.1 |
Clariom D TC |