Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000547420.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000547420.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000547420.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000547420.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000547420.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000547420.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
ENST00000552849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002441.hg.1 |
ENST00000216807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002441.hg.1 |
ENST00000543183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002441.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002441.hg.1 |
TC14000225.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000547420.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000547420.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000547420.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000547420.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000547420.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000547420.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
ENST00000552849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006913.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148059.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148059.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148059.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148059.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148059.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148059.hg.1 |
ENST00000216807.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148059.hg.1 |
ENST00000548758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148059.hg.1 |
ENST00000551774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148059.hg.1 |
ENST00000552677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148059.hg.1 |
NM_032352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148059.hg.1 |
TC1400006913.hg.1 |
Clariom D TC |