Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000544881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000544881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
ENST00000544881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016632.hg.1 |
ENST00000266397.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016632.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016632.hg.1 |
ENST00000544881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016632.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016632.hg.1 |
TC12001277.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200175712.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175712.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175712.hg.1 |
ENST00000544881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175712.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175712.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175712.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175712.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175712.hg.1 |
ENST00000544881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175712.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175712.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175712.hg.1 |
TC1200010023.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000266397.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000544881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000544881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
ENST00000544881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010023.hg.1 |
NM_152321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175715.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175715.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175715.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175715.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175715.hg.1 |
TC1200010023.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200175716.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175716.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175716.hg.1 |
ENST00000540097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175716.hg.1 |
NM_001300784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175716.hg.1 |
TC1200010023.hg.1 |
Clariom D TC |